Assay Details
| Gene Symbol: UBASH3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261286.1 | NP_001248215.1 | 1770 | 77 | ||
| XM_015115904.2 | XP_014971390.1 | 1579 | 77 | |||
| XM_015115905.2 | XP_014971391.1 | 2429 | 77 | |||
| GenBank mRNA | JU477558.1 | - | 1770 | 77 | ||
More Information
Set Membership: |
|

UBASH3B
Genomic Map