Assay Details
| Gene Symbol: PSMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261552.2 | NP_001248481.1 | 354 | 85 | ||
| XM_028832705.1 | XP_028688538.1 | 221 | 85 | |||
| GenBank mRNA | JU335558.1 | - | 349 | 85 | ||
| JU473782.1 | - | 347 | 85 | |||
| JV043164.1 | - | 354 | 85 | |||
| JV601922.1 | - | 355 | 85 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PSMC3
Genomic Map