Assay Details
| Gene Symbol: RIPOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148784.2 | XP_015004270.2 | 1620 | 109 | ||
| XM_028828505.1 | XP_028684338.1 | 1653 | 109 | |||
| XM_028828506.1 | XP_028684339.1 | 1508 | 109 | |||
| XM_028828507.1 | XP_028684340.1 | 1109 | 109 | |||
| XM_028828508.1 | XP_028684341.1 | 1575 | 109 | |||
| XM_028828509.1 | XP_028684342.1 | 1473 | 109 | |||
More Information
Set Membership: |
|

RIPOR3
Genomic Map