Assay Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193800.2 | NP_001180729.1 | 960 | 63 | ||
| GenBank mRNA | JU336993.1 | - | 907 | 63 | ||
| JU471293.1 | - | 964 | 63 | |||
| JV074930.1 | - | 812 | 63 | |||
More Information
Set Membership: |
|

OTULIN
Genomic Map