Assay Details
| Gene Symbol: RAB2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193860.2 | NP_001180789.1 | 346 | 78 | ||
| XM_015145400.2 | XP_015000886.1 | 615 | 78 | |||
| XM_028852147.1 | XP_028707980.1 | 615 | 78 | |||
| GenBank mRNA | JU319780.1 | - | 481 | 78 | ||
| JU470671.1 | - | 389 | 78 | |||
| JV047810.1 | - | 481 | 78 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAB2A
Genomic Map