Assay Details
| Gene Symbol: PLPPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122935.2 | XP_014978421.1 | 1408 | 61 | ||
| XM_015122936.2 | XP_014978422.1 | 1333 | 61 | |||
| XM_015122937.2 | XP_014978423.1 | 1392 | 61 | |||
| GenBank mRNA | JU335941.1 | - | 1377 | 61 | ||
| JU335942.1 | - | 1361 | 61 | |||
| JV044697.1 | - | 1358 | 61 | |||
| JV047948.1 | - | 1374 | 61 | |||
| JV047949.1 | - | 1283 | 61 | |||
| JV693476.1 | - | 1375 | 61 | |||
More Information
Set Membership: |
|

PLPPR2
Genomic Map