Assay Details
| Gene Symbol: CAMLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202851.1 | NP_001189780.1 | 712 | 71 | ||
| XM_015140861.2 | XP_014996347.1 | 1294 | 71 | |||
| XM_015140862.2 | XP_014996348.1 | 1294 | 71 | |||
| GenBank mRNA | JU336573.1 | - | 258 | 71 | ||
| JU336574.1 | - | 258 | 71 | |||
| JU336575.1 | - | 258 | 71 | |||
| JU336576.1 | - | 258 | 71 | |||
| JU477382.1 | - | 217 | 71 | |||
| JV048363.1 | - | 279 | 71 | |||
| JV559382.1 | - | 250 | 71 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CAMLG
Genomic Map