Assay Details
| Gene Symbol: TNFAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257463.1 | NP_001244392.1 | 722 | 82 | ||
| GenBank mRNA | JU336155.1 | - | 722 | 82 | ||
| JU477012.1 | - | 639 | 82 | |||
| JV048101.1 | - | 721 | 82 | |||
| JV592263.1 | - | 724 | 82 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TNFAIP1
Genomic Map