Assay Details
| Gene Symbol: ADNP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122249.2 | XP_014977735.1 | 769 | 110 | ||
| XM_015122250.2 | XP_014977736.1 | 556 | 110 | |||
| XM_015122251.2 | XP_014977737.1 | 198 | 110 | |||
| XM_015122252.2 | XP_014977738.1 | 170 | 110 | |||
| XM_028837904.1 | XP_028693737.1 | 1101 | 110 | |||
| XM_028837905.1 | XP_028693738.1 | 981 | 110 | |||
| GenBank mRNA | JU324312.1 | - | 228 | 110 | ||
| JU474716.1 | - | 127 | 110 | |||
| JV709779.1 | - | 220 | 110 | |||
More Information
Set Membership: |
|

ADNP2
Genomic Map