Assay Details
| Gene Symbol: ELK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261604.1 | NP_001248533.1 | 1405 | 61 | ||
| XM_015118422.2 | XP_014973908.1 | 1292 | 61 | |||
| GenBank mRNA | JU333301.1 | - | 1349 | 61 | ||
| JU474615.1 | - | 1405 | 61 | |||
More Information
Set Membership: |
|

ELK4
Genomic Map