Assay Details
| Gene Symbol: ITGB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265993.1 | NP_001252922.1 | 2719 | 122 | ||
| GenBank mRNA | JU334390.1 | - | 2719 | 122 | ||
| JV704357.1 | - | 2720 | 122 | |||
More Information
Set Membership: |
|

ITGB8
Genomic Map