Assay Details
| Gene Symbol: SLC41A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261258.1 | NP_001248187.1 | 601 | 73 | ||
| XM_015129935.2 | XP_014985421.1 | 738 | 73 | |||
| XM_015129936.2 | XP_014985422.1 | 586 | 73 | |||
| XM_015129937.2 | XP_014985423.1 | 525 | 73 | |||
| XM_028843430.1 | XP_028699263.1 | 613 | 73 | |||
| XM_028843431.1 | XP_028699264.1 | 714 | 73 | |||
| XM_028843432.1 | XP_028699265.1 | 554 | 73 | |||
| GenBank mRNA | JU322758.1 | - | 498 | 73 | ||
| JV044691.1 | - | 493 | 73 | |||
| JV044692.1 | - | 601 | 73 | |||
| JV636241.1 | - | 598 | 73 | |||
| JV638145.1 | - | 530 | 73 | |||
| JV669639.1 | - | 524 | 73 | |||
More Information
Set Membership: |
|

SLC41A3
Genomic Map