Assay Details
| Gene Symbol: CMSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193902.1 | NP_001180831.1 | 897 | 77 | ||
| XM_015128690.2 | XP_014984176.1 | 1829 | 77 | |||
| XM_028843159.1 | XP_028698992.1 | 777 | 77 | |||
| GenBank mRNA | JU323126.1 | - | 858 | 77 | ||
| JV048443.1 | - | 859 | 77 | |||
| JV489176.1 | - | 873 | 77 | |||
| JV636945.1 | - | 876 | 77 | |||
| JV703927.1 | - | 878 | 77 | |||
More Information
Set Membership: |
|

CMSS1
Genomic Map