Assay Details
| Gene Symbol: NKX2-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094853.4 | XP_001094853.1 | 864 | 147 | ||
| XM_028828130.1 | XP_028683963.1 | 820 | 147 | |||
| GenBank mRNA | JU328101.1 | - | 250 | 147 | ||
More Information
Set Membership: |
|

NKX2-2
Genomic Map