Assay Details
| Gene Symbol: TRIM36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266479.1 | NP_001253408.1 | 2050 | 66 | ||
| XM_015140570.2 | XP_014996056.1 | 2468 | 66 | |||
| GenBank mRNA | JU321497.1 | - | 2050 | 66 | ||
| JU321499.1 | - | 2050 | 66 | |||
More Information
Set Membership: |
|

TRIM36
Genomic Map