Assay Details
| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087145.3 | XP_001087145.1 | 531 | 79 | ||
| XM_015127494.2 | XP_014982980.1 | 737 | 79 | |||
| XM_015127496.2 | XP_014982982.1 | 756 | 79 | |||
| XM_015127498.2 | XP_014982984.1 | 665 | 79 | |||
| XM_028842141.1 | XP_028697974.1 | 591 | 79 | |||
| XM_028842142.1 | XP_028697975.1 | 500 | 79 | |||
| XM_028842143.1 | XP_028697976.1 | 242 | 79 | |||
| XM_028842144.1 | XP_028697977.1 | 534 | 79 | |||
| GenBank mRNA | JU323983.1 | - | 315 | 79 | ||
| JV053163.1 | - | 310 | 79 | |||
More Information
Set Membership: |
|

RIBC1
Genomic Map