Assay Details
| Gene Symbol: CYP17A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040232.1 | NP_001035322.1 | 439 | 70 | ||
| GenBank mRNA | AF458332.1 | - | 433 | 70 | ||
| AY746983.1 | - | 439 | 70 | |||
| JV044031.1 | - | 676 | 70 | |||
| JV643653.1 | - | 408 | 70 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CYP17A1
Genomic Map