Assay Details
| Gene Symbol: SLC25A30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121015.2 | XP_014976501.2 | 600 | 139 | ||
| XM_028837280.1 | XP_028693113.1 | 469 | 139 | |||
| XM_028837281.1 | XP_028693114.1 | 480 | 139 | |||
| XM_028837282.1 | XP_028693115.1 | 486 | 139 | |||
| GenBank mRNA | JU333780.1 | - | 420 | 139 | ||
| JV046641.1 | - | 316 | 139 | |||
| JV647301.1 | - | 440 | 139 | |||
More Information
Set Membership: |
|

SLC25A30
Genomic Map