Assay Details
| Gene Symbol: NR2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265967.2 | NP_001252896.1 | 1057 | 62 | ||
| GenBank mRNA | JU336146.1 | - | 1059 | 62 | ||
| JU336147.1 | - | 1059 | 62 | |||
More Information
Set Membership: |
|

NR2E1
Genomic Map