Assay Details
| Gene Symbol: NOXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082129.4 | XP_001082129.2 | 3174 | 103 | ||
| XM_015125335.2 | XP_014980821.1 | 817 | 103 | |||
| XM_015125336.2 | XP_014980822.1 | 3186 | 103 | |||
| XM_015125337.2 | XP_014980823.1 | 3171 | 103 | |||
| XM_028841031.1 | XP_028696864.1 | 3189 | 103 | |||
More Information
Set Membership: |
|

NOXO1
Genomic Map