Assay Details
| Gene Symbol: ZNF704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260976.1 | NP_001247905.1 | 578 | 80 | ||
| XM_015145518.2 | XP_015001004.2 | 1520 | 80 | |||
| XM_015145519.2 | XP_015001005.1 | 861 | 80 | |||
| GenBank mRNA | JU335824.1 | - | 580 | 80 | ||
| JU476701.1 | - | 578 | 80 | |||
More Information
Set Membership: |
|

ZNF704
Genomic Map