Assay Details
| Gene Symbol: TRIM39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135874.2 | XP_014991360.1 | 478 | 90 | ||
| XM_015135875.2 | XP_014991361.1 | 331 | 90 | |||
| XM_015135876.2 | XP_014991362.1 | 493 | 90 | |||
| XM_015135878.2 | XP_014991364.1 | 584 | 90 | |||
| XM_015135879.2 | XP_014991365.1 | 473 | 90 | |||
| XM_028846903.1 | XP_028702736.1 | 604 | 90 | |||
| GenBank mRNA | JU324789.1 | - | 177 | 90 | ||
| JV045345.1 | - | 128 | 90 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015135874.2, XM_015135875.2, XM_015135876.2, XM_015135878.2, XM_015135879.2, XM_028846903.1 transcripts.
Set Membership: |
|

TRIM39
Genomic Map