Assay Details
| Gene Symbol: TRIM39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135874.2 | XP_014991360.1 | 1410 | 88 | ||
| XM_015135875.2 | XP_014991361.1 | 1263 | 88 | |||
| XM_015135876.2 | XP_014991362.1 | 1425 | 88 | |||
| XM_015135878.2 | XP_014991364.1 | 1516 | 88 | |||
| XM_015135879.2 | XP_014991365.1 | 1405 | 88 | |||
| XM_028846903.1 | XP_028702736.1 | 1536 | 88 | |||
| GenBank mRNA | JU324789.1 | - | 1109 | 88 | ||
| JV045345.1 | - | 1060 | 88 | |||
More Information
Set Membership: |
|

TRIM39
Genomic Map