Assay Details
Gene Symbol: NOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258197.1 | NP_001245126.1 | 835 | 69 | ||
GenBank mRNA | JU323238.1 | - | 826 | 69 | ||
JU472754.1 | - | 835 | 69 | |||
JV504026.1 | - | 58 | 69 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
