Assay Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193777.2 | NP_001180706.1 | 713 | 78 | ||
| GenBank mRNA | JU320843.1 | - | 681 | 78 | ||
| JU473650.1 | - | 640 | 78 | |||
| JV044150.1 | - | 633 | 78 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM248
Genomic Map