Assay Details
| Gene Symbol: CXCL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193507.1 | NP_001180436.1 | 377 | 66 | ||
| GenBank mRNA | JU320880.1 | - | 327 | 66 | ||
| JU473660.1 | - | 308 | 66 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CXCL14
Genomic Map