Assay Details
| Gene Symbol: PCYOX1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266159.2 | NP_001253088.1 | 840 | 87 | ||
| XM_028849251.1 | XP_028705084.1 | 1501 | 87 | |||
| GenBank mRNA | JV593240.1 | - | 873 | 87 | ||
| JV636370.1 | - | 841 | 87 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PCYOX1L
Genomic Map