Assay Details
| Gene Symbol: SLC35E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266345.1 | NP_001253274.1 | 650 | 68 | ||
| GenBank mRNA | JU328471.1 | - | 335 | 68 | ||
| JU477267.1 | - | 645 | 68 | |||
| JU477268.1 | - | 645 | 68 | |||
| JU477269.1 | - | 645 | 68 | |||
| JU477270.1 | - | 645 | 68 | |||
| JU477271.1 | - | 645 | 68 | |||
| JV048292.1 | - | 650 | 68 | |||
More Information
Set Membership: |
|

SLC35E1
Genomic Map