Assay Details
| Gene Symbol: SLC25A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258165.1 | NP_001245094.1 | 723 | 86 | ||
| GenBank mRNA | JU334161.1 | - | 676 | 86 | ||
| JU475287.1 | - | 723 | 86 | |||
| JV046845.1 | - | 718 | 86 | |||
| JV649356.1 | - | 724 | 86 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC25A4
Genomic Map