Assay Details
| Gene Symbol: SLPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168663.1 | NP_001162134.1 | 411 | 91 | ||
| GenBank mRNA | CO579827.1 | - | 394 | 91 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLPI
Genomic Map