Assay Details
| Gene Symbol: FDFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260724.1 | NP_001247653.1 | 283 | 83 | ||
| XM_015144737.2 | XP_015000223.1 | 423 | 83 | |||
| GenBank mRNA | JU322334.1 | - | 340 | 83 | ||
| JU474700.1 | - | 282 | 83 | |||
| JV046458.1 | - | 283 | 83 | |||
| JV669076.1 | - | 297 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FDFT1
Genomic Map