Assay Details
| Gene Symbol: MRPL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266484.1 | NP_001253413.1 | 308 | 65 | ||
| GenBank mRNA | JV047869.1 | - | 308 | 65 | ||
| JV602493.1 | - | 389 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRPL23
Genomic Map