Assay Details
| Gene Symbol: IFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032905.1 | NP_001028077.1 | 187 | 150 | ||
| GenBank mRNA | AY376145.1 | - | 187 | 150 | ||
| L26024.1 | - | 187 | 150 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IFNG
Genomic Map