Assay Details
| Gene Symbol: CSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032949.2 | NP_001028121.1 | 219 | 96 | ||
| GenBank mRNA | AY007376.1 | - | 219 | 96 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CSF2
Genomic Map