Assay Details
| Gene Symbol: CCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032821.1 | NP_001027993.1 | 77 | 104 | ||
| GenBank mRNA | AF255343.1 | - | 77 | 104 | ||
| AF449265.1 | - | 77 | 104 | |||
| JU346953.1 | - | 130 | 104 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CCL2
Genomic Map