Assay Details
| Gene Symbol: STEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134141.2 | XP_014989627.1 | 1718 | 153 | ||
| XM_015134142.2 | XP_014989628.2 | 1401 | 153 | |||
| XM_015134144.2 | XP_014989630.1 | 1503 | 153 | |||
| XM_028846090.1 | XP_028701923.1 | 1502 | 153 | |||
| XM_028846091.1 | XP_028701924.1 | 1392 | 153 | |||
| XM_028846092.1 | XP_028701925.1 | 1509 | 153 | |||
| XM_028846093.1 | XP_028701926.1 | 1504 | 153 | |||
| XM_028846094.1 | XP_028701927.1 | 1508 | 153 | |||
| GenBank mRNA | JU321585.1 | - | 1167 | 153 | ||
| JV704194.1 | - | 1230 | 153 | |||
More Information
Set Membership: |
|

STEAP2
Genomic Map