Assay Details
| Gene Symbol: MIEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194095.3 | NP_001181024.1 | 557 | 143 | ||
| XM_001098384.4 | XP_001098384.1 | 730 | 143 | |||
| GenBank mRNA | JU321307.1 | - | 558 | 143 | ||
| JU473451.1 | - | 503 | 143 | |||
| JV584979.1 | - | 619 | 143 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MIEF1
Genomic Map