Assay Details
| Gene Symbol: DEFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032857.1 | NP_001028029.1 | 77 | 78 | ||
| GenBank mRNA | AF014016.1 | - | 77 | 78 | ||
| AF288285.1 | - | 77 | 78 | |||
More Information
Set Membership: |
|

DEFB1
Genomic Map