Assay Details
| Gene Symbol: GPR15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042647.1 | NP_001036112.1 | 50 | 80 | ||
| GenBank mRNA | AF084226.1 | - | 50 | 80 | ||
| AF084227.1 | - | 50 | 80 | |||
| DQ887987.1 | - | 50 | 80 | |||
| DQ887992.1 | - | 50 | 80 | |||
| DQ887993.1 | - | 50 | 80 | |||
| DQ887994.1 | - | 50 | 80 | |||
| DQ887996.1 | - | 50 | 80 | |||
| DQ887997.1 | - | 50 | 80 | |||
| DQ887998.1 | - | 50 | 80 | |||
| DQ887999.1 | - | 80 | ||||
| DQ888002.1 | - | 50 | 80 | |||
| DQ888003.1 | - | 50 | 80 | |||
| DQ888004.1 | - | 50 | 80 | |||
| DQ888006.1 | - | 50 | 80 | |||
| DQ888010.1 | - | 63 | 80 | |||
| DQ888012.1 | - | 50 | 80 | |||
| DQ888013.1 | - | 50 | 80 | |||
| DQ888016.1 | - | 50 | 80 | |||
| DQ888017.1 | - | 50 | 80 | |||
| DQ888020.1 | - | 54 | 80 | |||
| DQ888024.1 | - | 50 | 80 | |||
| DQ888025.1 | - | 50 | 80 | |||
| DQ888026.1 | - | 50 | 80 | |||
| DQ888027.1 | - | 50 | 80 | |||
| DQ888028.1 | - | 50 | 80 | |||
| DQ888029.1 | - | 50 | 80 | |||
| DQ888031.1 | - | 50 | 80 | |||
| DQ888035.1 | - | 50 | 80 | |||
| DQ888036.1 | - | 50 | 80 | |||
| DQ888037.1 | - | 50 | 80 | |||
| DQ888038.1 | - | 50 | 80 | |||
More Information
Set Membership: |
|

GPR15
Genomic Map