Assay Details
| Gene Symbol: TNFRSF13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118722.2 | XP_014974208.1 | 822 | 92 | ||
| XM_015118723.2 | XP_014974209.1 | 687 | 92 | |||
| XM_028836059.1 | XP_028691892.1 | 680 | 92 | |||
| GenBank mRNA | AF227558.1 | - | 354 | 92 | ||
More Information
Set Membership: |
|

TNFRSF13B
Genomic Map