Assay Details
| Gene Symbol: NPY5R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032833.1 | NP_001028005.1 | 1238 | 126 | ||
| XM_015139386.2 | XP_014994872.1 | 2268 | 126 | |||
| XM_015139387.2 | XP_014994873.1 | 2272 | 126 | |||
| XM_015139388.2 | XP_014994874.1 | 1565 | 126 | |||
| XM_015139389.2 | XP_014994875.1 | 2271 | 126 | |||
| XM_028847924.1 | XP_028703757.1 | 2140 | 126 | |||
| XM_028847925.1 | XP_028703758.1 | 2143 | 126 | |||
| GenBank mRNA | AF303091.1 | - | 1238 | 126 | ||
More Information
Set Membership: |
|

NPY5R
Genomic Map