Assay Details
| Gene Symbol: PI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110511.1 | NP_001103981.1 | 142 | 69 | ||
| GenBank mRNA | AJ223215.1 | - | 51 | 69 | ||
| JU473420.1 | - | 84 | 69 | |||
More Information
Set Membership: |
|

PI3
Genomic Map