Assay Details
| Gene Symbol: MAMU-DPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048247.1 | NP_001041712.1 | 346 | 68 | ||
| GenBank mRNA | AB219099.1 | - | 346 | 68 | ||
| EF204948.1 | - | 346 | 68 | |||
| HM594293.1 | - | 346 | 68 | |||
More Information
Set Membership: |
|

MAMU-DPA
Genomic Map