Assay Details
| Gene Symbol: GYPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139289.2 | XP_014994775.2 | 409 | 74 | ||
| XM_015139291.2 | XP_014994777.2 | 286 | 74 | |||
| XM_028848783.1 | XP_028704616.1 | 497 | 74 | |||
| GenBank mRNA | AF023469.1 | - | 236 | 74 | ||
More Information
Set Membership: |
|

GYPA
Genomic Map