Assay Details
| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032891.1 | NP_001028063.1 | 632 | 58 | ||
| XM_015124855.2 | XP_014980341.2 | 885 | 58 | |||
| XM_028838326.1 | XP_028694159.1 | 851 | 58 | |||
| GenBank mRNA | AY035218.1 | - | 632 | 58 | ||
| AY035219.1 | - | 632 | 58 | |||
| AY035220.1 | - | 632 | 58 | |||
| AY035221.1 | - | 632 | 58 | |||
| AY035222.1 | - | 596 | 58 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NCR1
Genomic Map