Assay Details
| Gene Symbol: LOC705095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133688.2 | XP_014989174.1 | 713 | 250 | ||
| XM_028845859.1 | XP_028701692.1 | 828 | 250 | |||
More Information
Set Membership: |
|

LOC705095
Genomic Map