Assay Details
| Gene Symbol: NLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118887.2 | XP_014974373.1 | 2589 | 112 | ||
| XM_015118888.2 | XP_014974374.1 | 2504 | 112 | |||
| GenBank mRNA | AY680464.1 | - | 264 | 112 | ||
| JU320404.1 | - | 1642 | 112 | |||
| JV637368.1 | - | 1651 | 112 | |||
More Information
Set Membership: |
|

NLK
Genomic Map