Assay Details
| Gene Symbol: AKAP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278476.1 | NP_001265405.1 | 6036 | 102 | ||
| NM_001278477.1 | NP_001265406.1 | 5744 | 102 | |||
| XM_015137072.2 | XP_014992558.2 | 6009 | 102 | |||
| XM_015137073.2 | XP_014992559.2 | 5810 | 102 | |||
| XM_015137074.2 | XP_014992560.2 | 5589 | 102 | |||
| XM_015137075.2 | XP_014992561.2 | 5626 | 102 | |||
| GenBank mRNA | AY680469.1 | - | 504 | 102 | ||
| JU337231.1 | - | 5945 | 102 | |||
| JU337232.1 | - | 5945 | 102 | |||
| JV048786.1 | - | 5678 | 102 | |||
| JV599119.1 | - | 5983 | 102 | |||
| JV634215.1 | - | 5982 | 102 | |||
| JV634216.1 | - | 5744 | 102 | |||
| JV634217.1 | - | 5744 | 102 | |||
| JV666269.1 | - | 6036 | 102 | |||
| JV727516.1 | - | 5980 | 102 | |||
More Information
Set Membership: |
|

AKAP12
Genomic Map