Assay Details
| Gene Symbol: ZYG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028833745.1 | XP_028689578.1 | 2249 | 75 | ||
| GenBank mRNA | DQ148057.1 | - | 179 | 75 | ||
| DQ148070.1 | - | 132 | 75 | |||
| JU320581.1 | - | 2159 | 75 | |||
| JU320582.1 | - | 2159 | 75 | |||
| JU477224.1 | - | 2209 | 75 | |||
| JV048263.1 | - | 2083 | 75 | |||
More Information
Set Membership: |
|

ZYG11B
Genomic Map