Assay Details
| Gene Symbol: CXCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032965.1 | NP_001028137.1 | 254 | 65 | ||
| GenBank mRNA | S78555.1 | - | 204 | 65 | ||
| U19849.1 | - | 254 | 65 | |||
More Information
Set Membership: |
|

CXCL8
Genomic Map